missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Emerin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529731
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HeLa nucleus. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Emerin Polyclonal antibody specifically detects Emerin in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Emerin | |
Polyclonal | |
Unconjugated | |
Emd | |
AW550900; EDMD; EMD; Emerin; LEM domain containing 5; LEMD5; STA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2010 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P50402 | |
Emd | |
Recombinant fragment corresponding to a region within amino acids 48 and 254 of Human Emerin | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |