missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA533502
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon (93%) Monkey (87%) Marmoset, Mouse, Rat, Pig, Platypus (80%).
EMR1 Polyclonal antibody specifically detects EMR1 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
EMR1 | |
Polyclonal | |
Unconjugated | |
ADGRE1 | |
ADGRE1; Adhesion G protein-coupled receptor E1; Adhesion G protein-coupled receptor E2; cell surface glycoprotein EMR1; cell surface glycoprotein F4/80; DD7A5-7; egf-like module containing, mucin-like, hormone receptor-like 1; EGF-like module containing, mucin-like, hormone receptor-like sequence 1; EGF-like module receptor 1; EGF-like module-containing mucin-like hormone receptor-like 1; EGF-TM7; Emr1; EMR1 hormone receptor; F4/80; Gpf480; Ly71; lymphocyte antigen 71; TM7LN3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2015 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q14246 | |
ADGRE1 | |
Synthetic 16 amino acid peptide from N-terminal extracellular domain of human EMR1. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |