missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMR2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA533507
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Monkey (90%).
EMR2 Polyclonal antibody specifically detects EMR2 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
EMR2 | |
Polyclonal | |
Unconjugated | |
ADGRE2 | |
ADGRE2; Adhesion G protein-coupled receptor E2; CD312; CD312 antigen; egf-like module containing, mucin-like, hormone receptor-like 2; EGF-like module receptor 2; EGF-like module-containing mucin-like hormone receptor-like 2; EMR2; VBU | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
30817 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9UHX3 | |
ADGRE2 | |
Synthetic 20 amino acid peptide from N-terminal extracellular domain of human EMR2. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |