missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMX2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA128062
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-28062 detects EMX2 from human, mouse, rat samples.
EMX2 Polyclonal antibody specifically detects EMX2 in Human, Rodent samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
EMX2 | |
Polyclonal | |
Unconjugated | |
EMX2 | |
empty spiracles homeobox 2; empty spiracles homolog 2; Empty spiracles-like protein 2; Emx2; Emx-2; Homeobox protein EMX2; Pdo; RGD1564797 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2018 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% ammonium sulfate and no preservative | |
Q04743 | |
EMX2 | |
synthetic peptide PSSHSPHPLFASQQRDPS conjugated to BSA, corresponding to amino acids 100-117 of Human EMX2. | |
50 μg | |
Primary | |
Human, Rodent | |
Antibody | |
IgG |