missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EN1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA514149
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
EN1 Polyclonal antibody specifically detects EN1 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
EN1 | |
Polyclonal | |
Unconjugated | |
EN1 | |
En1; En-1; engrailed 1; engrailed homeobox 1; engrailed homolog 1; engrailed-1; HME1; homeobox protein en-1; Homeobox protein engrailed-1; hu-En-1; Mo-en.1; mo-En-1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
13798, 2019 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P09065, Q05925 | |
EN1 | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human EN1 (Engrailed 1) | |
200 μL | |
Primary | |
Human, Mouse | |
Antibody | |
Ig |