Learn More
Nidogen G1 Domain Monoclonal Antibody (JF3), Invitrogen™
Rat Monoclonal Antibody
Marke: Invitrogen MA517019
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-17019 recognizes native and denatured mouse and human nidogen, and blocks binding of nidogen to laminin. MA5-17019 has been successfully used in ELISA and Western blot applications. The MA5-17019 immunogen is a recombinant fragment corresponding to the G1Link region of mouse nidogen produced in Fisher rat plus YB2/0 cells.
Nidogen G1 Domain Monoclonal antibody specifically detects Nidogen G1 Domain in Human, Mouse samples. It is validated for ELISA, Western Blot.Spezifikation
Nidogen G1 Domain | |
Monoclonal | |
1 mg/mL | |
2M acetic acid, 2M tris with no preservative | |
P10493, P14543 | |
NID1 | |
Recombinant fragment corresponding to the G1Link region of mouse nidogen produced in Fisher rat plus YB2/0 cells | |
50 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a |
ELISA, Western Blot | |
JF3 | |
Unconjugated | |
NID1 | |
A630025O17; enactin; Ent; Entactin; entactin 1; entactin-1; NID; NID 1; Nid1; NID-1; Nidogen (enactin); nidogen 1; Nidogen precursor (Entactin); nidogen-1 | |
Rat | |
Affinity chromatography | |
RUO | |
18073, 4811 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |