missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENDOG Monoclonal Antibody (7F2D7), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA515758
Weitere Details : Netto-Gewicht : 0.10000kg
Beschreibung
A suggested positive control is HepG2 cell lysate.
ENDOG Monoclonal antibody specifically detects ENDOG in Human, Mouse, Rat samples. It is validated for Western Blot.Spezifikation
ENDOG | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O08600, Q14249 | |
ENDOG | |
Recombinant protein corresponding to amino acids 51 - 140 of human EndoG. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Western Blot | |
7F2D7 | |
Unconjugated | |
ENDOG | |
endo G; Endog; endonuclease G; endonuclease G, mitochondrial; mitochondrial endonuclease G | |
Mouse | |
Protein A | |
RUO | |
13804, 2021, 362100 | |
-20°C | |
Liquid |