missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENDOGL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA539161
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
This gene encodes an endo/exonuclease with 5′-3′ exonuclease activity. The encoded enzyme catalyzes the hydrolysis of ester linkages at the 5′ end of a nucleic acid chain. This enzyme is localized to the mitochondria and may play a role in programmed cell death. Alternatively spliced transcript variants have been described. A pseudogene exists on chromosome 18.Spezifikation
ENDOGL1 | |
Polyclonal | |
Unconjugated | |
EXOG | |
Nuclease EXOG, mitochondrial, Endonuclease G-like 1, Endo G-like 1, EXOG, ENGL, ENDOGL2, ENDOGL1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9941 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9Y2C4 | |
EXOG | |
A synthetic peptide derived from the internal region of human ENDOGL1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |