missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNDC5 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518297
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
TXNDC5 Polyclonal antibody specifically detects TXNDC5 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Western Blot.Spezifikation
TXNDC5 | |
Polyclonal | |
Unconjugated | |
Txndc5 | |
AL022641; ENDOPDI; endoplasmic reticulum protein ERp46; endoplasmic reticulum resident protein 46; endothelial protein disulphide isomerase; ER protein 46; ERp46; FLJ21353; FLJ90810; HCC2; HCC-2; MGC3178; PC-TRP; PDIA15; Plasma cell-specific thioredoxin-related protein; protein disulfide isomerase family A, member 15; STRF8; thiore; thioredoxin domain containing 5; thioredoxin domain containing 5 (endoplasmic reticulum); thioredoxin domain-containing protein 5; thioredoxin related protein; thioredoxin-like protein p46; Tlp46; TXNDC5; UNQ364; UNQ364/PRO700 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
81567 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q8NBS9 | |
Txndc5 | |
Synthetic peptide sequence (SLHRFVLSQAKDEL) corresponding to the C-terminus amino acids of TXNDC5 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |