missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RECA-1 Monoclonal Antibody (HIS-52), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA135781
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
RECA-1 is a cell surface antigen which is expressed by all rat endothelial cells. The endothelium is the thin layer of cells that line the interior surface of blood vessels, forming an interface between circulating blood in the lumen and the rest of the vessel wall. Endothelial cells line the entire circulatory system, from the heart to the smallest capillary.Spezifikation
RECA-1 | |
Monoclonal | |
Unconjugated | |
Mouse | |
1 mL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot | |
HIS-52 | |
Endothelial cells; Endothelium Cell Antigen-1; Endothelial cell marker; Pan endothelial cells | |
Peripheral and mesenteric lymph nodes isolate. | |
RUO | |
Rat | |
Antibody | |
IgG1 |