missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIPG Polyclonal Antibody, Biotin, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA522916
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody does not react with mouse endothelial lipase.
LIPG Polyclonal antibody specifically detects LIPG in Human samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Spezifikation
LIPG | |
Polyclonal | |
Biotin | |
LIPG | |
3110013K01Rik; EDL; EL; endothelial cell-derived lipase; Endothelial lipase; endothelial-derived lipase; lipase; lipase G; lipase G, endothelial type; lipase, endothelial; LIPG; lipoprotein lipase H; lipose, endothelial; mEDL; PL; PRO719; UNQ387/PRO719 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9388 | |
4° C, store in dark | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
0.9 mg/mL | |
PBS with 0.05% sodium azide | |
Q9Y5X9 | |
LIPG | |
An N-terminal synthetic peptide made to the human endothelial lipase protein sequence. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |