missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIPG Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA519032
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
LIPG Polyclonal antibody specifically detects LIPG in Human samples. It is validated for Western Blot.Spezifikation
LIPG | |
Polyclonal | |
Unconjugated | |
LIPG | |
3110013K01Rik; EDL; EL; endothelial cell-derived lipase; Endothelial lipase; endothelial-derived lipase; lipase; lipase G; lipase G, endothelial type; lipase, endothelial; LIPG; lipoprotein lipase H; lipose, endothelial; mEDL; PL; PRO719; UNQ387/PRO719 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
9388 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9Y5X9 | |
LIPG | |
Synthetic peptide sequence (RFNLRTSKDPEHEG) corresponding to the internal amino acids of LIPG | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |