missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endothelium Monoclonal Antibody (OX-43), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA170023
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
For extended storage, aliquot and store at -20oC or below. Avoid freeze-thaw cycles.
Endothelium Monoclonal antibody specifically detects Endothelium in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation.Spezifikation
Endothelium | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Mouse | |
Protein G | |
RUO | |
Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
OX-43 | |
Unconjugated | |
Endothelial cell marker; Endothelial cells; Endothelium Cell Antigen-1; Pan endothelial cells | |
Rat peritoneal macrophages. | |
250 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |