missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endothelium Monoclonal Antibody (OX-43), PE, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA517518
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-17518 targets Endothelium in FACS applications and shows reactivity with Rat samples. The MA5-17518 immunogen is rat Peritoneal Macrophages.
Endothelium Monoclonal antibody specifically detects Endothelium in Rat samples. It is validated for Flow Cytometry.Spezifikation
Endothelium | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
Mouse | |
Protein G | |
RUO | |
Rat | |
Antibody | |
IgG1 |
Flow Cytometry | |
OX-43 | |
PE | |
Endothelial cell marker; Endothelial cells; Endothelium Cell Antigen-1; Pan endothelial cells | |
Rat Peritoneal Macrophages | |
50 μg | |
Primary | |
4° C, do not freeze | |
Liquid |