missing translation for 'onlineSavingsMsg'
Learn More

NSE Monoclonal Antibody (NSE-P2), Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA517628

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 15383911

  • 419.00 € / 200 Mikrogramm
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

MA5-17628 targets ENO2 in ELISA, IHC, and WB applications and shows reactivity with Human samples. The MA5-17628 immunogen is enolase of human origin.

NSE Monoclonal antibody specifically detects NSE in Human samples. It is validated for ELISA, Immunohistochemistry, Western Blot.
Spezifikation

Spezifikation

NSE
Monoclonal
1 mg/mL
PBS with 0.02% sodium azide
P09104
ENO2
Enolase of human origin
200 μg
Primary
Human
Antibody
IgG1
ELISA, Immunohistochemistry, Western Blot
NSE-P2
Unconjugated
ENO2
2-phospho-D-glycerate hydrolyase; 2-phospho-D-glycerate hydro-lyase; AI837106; D6Ertd375e; Eno; ENO2; Eno-2; ENOG; Enolase 2; enolase 2 (gamma, neuronal); enolase 2, gamma neuronal; enolase 2, gamma, neuronal; epididymis secretory protein Li 279; gamma enolase; gamma-enolase; gamma-enolase-like; gamma-subunit of enolase; HEL-S-279; LOC100911625; Neural enolase; neuron specific enolase; neuron specific gamma enolase; Neuronal Enolase; neuronal enriched enolase; neurone-specific enolase; neuron-specific enolase; NSE; RNEN3
Mouse
Protein G
RUO
2026
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications
Sonderangebote

Sonderangebote