Learn More
eNOS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA516887
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA5-16887 targets eNOS in immunohistochemistry (paraffin) applications and shows reactivity with Bovine, Canine, Human, mouse, Porcine, and Rat samples. Suggested positive control is capillary endothelium and regenerative endothelium. The PA5-16887 immunogen is a synthetic peptide derived from C-ternimal of human eNOS.
eNOS Polyclonal antibody specifically detects eNOS in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
eNOS | |
Polyclonal | |
Unconjugated | |
NOS3 | |
2310065A03Rik; cNOS; Constitutive NOS; Ecnos; EC-NOS; endothelial nitric oxide synthase; endothelial nitric oxide synthase 3; endothelial nitric oxide synthase NOS3; endothelial NOS; eNos; e-NOS; nitric oxide synthase; nitric oxide synthase 3; nitric oxide synthase 3 (endothelial cell); nitric oxide synthase 3 transcript variant eNOS-delta20; nitric oxide synthase 3 transcript variant eNOS-delta20-21; nitric oxide synthase 3 transcript variant eNOS-delta21; nitric oxide synthase 3, endothelial cell; nitric oxide synthase, endothelial; NOS; NOS type III; Nos3; Nos-3; NOSIII; OTTHUMP00000213183 | |
Rabbit | |
Affinity chromatography | |
RUO | |
4846 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.081 mg/mL | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
P29474 | |
NOS3 | |
A synthetic peptide derived from C-ternimal of human eNOS | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG |