Learn More
ENOSF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531476
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Pig (88%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ENOSF1 Polyclonal antibody specifically detects ENOSF1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
ENOSF1 | |
Polyclonal | |
Unconjugated | |
ENOSF1 | |
antisense RNA to thymidylate synthase; enolase superfamily member 1; ENOSF1; HSRTSBETA; L-fuconate dehydratase; mitochondrial enolase superfamily member 1; rTS; rTS alpha; rTS beta; TYMSAS | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55556 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q7L5Y1 | |
ENOSF1 | |
Recombinant fragment corresponding to a region within amino acids 13 and 349 of Human ENOSF1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |