Learn More
Enterovirus VP-1 Polyclonal Antibody, Invitrogen™
Enterovirus VP-1 Antibody, PA5-32258, from Invitrogen™. Species Reactivity: Virus; Applications: ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry.
Marke: Invitrogen PA532258
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Enterovirus VP-1 Antibody, PA5-32258, from Invitrogen. Species Reactivity: Virus; Applications: ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry.
Host Species: Rabbit
Cross Reactivity: Virus
Isotype: IgG
Immunogen: Purified Enterovirus VP-1 antigen
Formulation: 100 ug purified antibody (2.5mg/mL) in PBS and 0.1% sodium azide; pH 7.4
Synonyms: Capsid VP-1
PA5-32258 targets Enterovirus VP-1 in ELISA and IF applications and shows reactivity with Virus samples. The PA5-32258 immunogen is purified Enterovirus VP-1 antigen.
Spezifikation
Enterovirus VP-1 | |
Polyclonal | |
Capsid VP-1 | |
Purified Enterovirus VP-1 antigen | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
100 μg | |
Virus | |
Purified |