missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENTPD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532751
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan, Gibbon, Monkey, Marmoset (100%) Galago, Hamster (80%).
ENTPD2 Polyclonal antibody specifically detects ENTPD2 in Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
ENTPD2 | |
Polyclonal | |
Unconjugated | |
ENTPD2 | |
CD39 antigen-like 1; CD39L1; Ecto-ATP diphosphohydrolase 2; ecto-ATPase 2; ecto-ATPDase 2; Ectonucleoside triphosphate diphosphohydrolase 2; ENTPD2; NTPDase 2; NTPDase2; NTPDase-2; testicular ecto-ATPase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
954 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9Y5L3 | |
ENTPD2 | |
Synthetic 15 amino acid peptide from internal region of human ENTPD2. | |
50 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |