missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p300 Monoclonal Antibody (NM-11), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen 337600
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
p300 Monoclonal antibody specifically detects p300 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.Spezifikation
p300 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
B2RWS6, Q09472 | |
EP300 | |
Recombinant human p300 protein | |
50 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
NM-11 | |
Unconjugated | |
EP300 | |
A430090G16; A730011L11; E1A binding protein p300; E1A-associated protein p300; E1A-binding protein, 300kD; Ep300; Histone acetyltransferase p300; Histone butyryltransferase p300; Histone crotonyltransferase p300; KAT3B; P300; p300 HAT; Protein propionyltransferase p300; RSTS2 | |
Mouse | |
Protein A | |
RUO | |
2033, 328572 | |
-20°C | |
Liquid |