missing translation for 'onlineSavingsMsg'
Learn More

estrogen receptor 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Artikelnummer. 16197527
Change view
Click to view available options
Menge:
100 μg
Packungsgröße:
100 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16197527 100 μg 100 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 16197527 Lieferant Abnova Lieferanten-Nr. H00002099D01P.100ug

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit polyclonal antibody raised against a full-length human ESR1 protein.

This gene encodes an estrogen receptor, a ligand-activated transcription factor composed of several domains important for hormone binding, DNA binding, and activation of transcription. The protein localizes to the nucleus where it may form a homodimer or a heterodimer with estrogen receptor 2. Estrogen and its receptors are essential for sexual development and reproductive function, but also play a role in other tissues such as bone. Estrogen receptors are also involved in pathological processes including breast cancer, endometrial cancer, and osteoporosis. Alternative splicing results in several transcript variants, which differ in their 5' UTRs and use different promoters. [provided by RefSeq]

Sequence: MTMTLHTKASGMALLHQIQGNELEPLNRPQLKIPLERPLGEVYLDSSKPAVYNYPEGAAYEFNAAAAANAQVYGQTGLPYGPGSEAAAFGSNGLGGFPPLNSVSPSPLMLLHPPPQLSPFLQPHGQQVPYYLENEPSGYTVREAGPPAFYRPNSDNRRQGGRERLASTNDKGSMAMESAKETRYCAVCNDYASGYHYGVWSCEGCKAFFKRSIQGHNDYMCPATNQCTIDKNRRKSCQACRLRKCYEVGMMKGGIRKDRRGGRMLKHKRQRDDGEGRGEVGSAGDMRAANLWPSPLMIKRSKKNSLALSLTADQMVSALLDAEPPILYSEYDPTRPFSEASMMGLLTNLADRELVHMINWAKRVPGFVDLTLHDQVHLLECAWLEILMIGLVWRSMEHPGKLLFAPNLLLDRNQGKCVEGMVEIFDMLLATSSRFRMMNLQGEEFVCLKSIILLNSGVYTFLSSTLKSLEEKDHIHRVLDKITDTLIHLMAKAGLTLQQQHQRLAQLLLILSHIRHMSNKGMEHLYSMKCKNVVPLYDLLLEMLDAHRLHAPTSRGGASVEETDQSHLATAGSTSSHSLQKYYITGEAEGFPATV

Spezifikation

Antigen estrogen receptor 1
Anwendungen Immunofluorescence, Western Blot
Klassifikation Polyclonal
Konjugat Unconjugated
Beschreibung Rabbit polyclonal antibody raised against a full-length human ESR1 protein.
Zusammensetzung PBS with no preservative; pH 7.4
Gen ESR1
Gen-Zugriffsnummer NM_000125.2
Gen-Alias DKFZp686N23123/ER/ESR/ESRA/Era/NR3A1
Gensymbole ESR1
Wirtsspezies Rabbit
Immunogen ESR1 (AAI28574.1, 1 a.a. ∼ 595 a.a) full-length human protein.
Reinigungsverfahren Affinity Purified
Menge 100 μg
Regulatorischer Status RUO
Forschungsgebiet Cell Cycle
Primär oder sekundär Primary
Gen-ID (Entrez) 2099
Zielspezies Human
Inhalt und Lagerung Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Produkttyp Antibody
Isotype IgG
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • ESR1 purified MaxPab rabbit polyclonal antibody (D01P)
Signalwort
  • Achtung
Gefahrenkategorie
  • Akute Toxizität Kategorie 4
  • Schwere Augenschädigung/Augenreizung Kategorie 2
  • Hautreizende/-ätzende Wirkung Kategorie 2
Gefahrenhinweis
  • H302-Gesundheitsschädlich bei Verschlucken.
  • H315-Verursacht Hautreizungen.
  • H319-Verursacht schwere Augenreizung.
Sicherheitshinweis
  • P102-Darf nicht in die Hände von Kindern gelangen.
  • P103-Vor Gebrauch Kennzeichnungsetikett lesen.
  • P233-Behälter dicht verschlossen halten.
  • P264-Nach Gebrauch gründlich waschen.
  • P270-Bei Gebrauch nicht essen, trinken oder rauchen.
  • P280-Schutzhandschuhe/Schutzkleidung/Augenschutz/Gesichtsschutz tragen.
  • P301+P310-BEI VERSCHLUCKEN: Sofort GIFTINFORMATIONSZENTRUM/Arzt anrufen/
  • P303+P361+P353-BEI BERÜHRUNG MIT DER HAUT (oder dem Haar): Alle kontaminierten Kleidungsstücke sofort ausziehen. Haut mit Wasser abwaschen/duschen.
  • P404-In einem geschlossenen Behälter aufbewahren.
  • P501b-Inhalt/Behälter gemäß den örtlichen/regionalen/nationalen/internationalen Vorschriften zuführen.
Ergänzende Informationen
  • MIXTURE LIST-Contain: sodium phosphate dibasic, potassium phosphate monobasic
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.