missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAIM Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518652
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with canine, mouse and rat based on sequence homology.
FAIM Polyclonal antibody specifically detects FAIM in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
FAIM | |
Polyclonal | |
Unconjugated | |
FAIM | |
Faim; FAIM 1; FAIM1; Fas apoptotic inhibitory molecule; fas apoptotic inhibitory molecule 1; rFAIM | |
Goat | |
Antigen affinity chromatography | |
RUO | |
55179 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9NVQ4 | |
FAIM | |
Synthetic peptide sequence (KKYMEDRSKTTN) corresponding to the internal amino acids of FAIM | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |