missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM47E Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA546681
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: KLEDTWAYCQ DTRKGMKEPT KLLKKHSTQV YLGPSKKTSV SNAGQWLYEE Sequence homology: Human: 100%; Mouse: 85%
FAM47E Polyclonal antibody specifically detects FAM47E in Human samples. It is validated for Western Blot.Spezifikation
FAM47E | |
Polyclonal | |
Unconjugated | |
FAM47E | |
C730043O17; FAM47E; family with sequence similarity 47 member E; family with sequence similarity 47, member E; Gm1381; Protein FAM47E | |
Rabbit | |
Affinity Chromatography | |
RUO | |
100129583 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6ZV65 | |
FAM47E | |
Synthetic peptide directed towards the middle region of human FAM47E | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |