missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FASTKD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA538810
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
This gene encodes a protein that is localized in the mitochondrial inner compartment and that may play a role in mitochondrial apoptosis. Nonsense mutations have been reported to result in cytochrome c oxidase deficiency. [provided by RefSeq, Oct 2008].Spezifikation
FASTKD2 | |
Polyclonal | |
Unconjugated | |
FASTKD2 | |
KIAA0971 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22868 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9NYY8 | |
FASTKD2 | |
A synthetic peptide derived from the internal region of human FAKD2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |