missing translation for 'onlineSavingsMsg'
Learn More

Foxp3 Rat anti-Mouse, Alexa Fluor 488, Clone: MF23, BD

Rat Monoclonal Antibody

Marke:  BD Biosciences 560407

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15800179

  • 331.00 € / 1 Stück
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Foxp3 is a 50-55kDa protein also known as Forkhead box P3, JM2, or IPEX. It is a member of the forkhead or winged helix family of transcription factors and is specifically expressed by T regulatory (Treg) cells. Foxp3 has been reported to be a key regulatory protein for Treg cell development and function. Ectopic expression of Foxp3 in conventional T cells is sufficient to induce suppressive activity, repress the production of cytokines such as IL2 and IFN-γ, and upregulate Treg cell-associated molecules such as CD25, CTLA4 and GITR. It has been found that the mutation of Foxp3 is responsible for “scurfy” mice. When overexpressed, Foxp3 leads to poor T cell proliferation and activation.
Monoclonal antibody MF23 reacts with mouse Foxp3. Immunoblotting with MF23 antibody has confirmed it recognizes an epitope between 1-87 amino acids in the N-terminal domain.

Host Species: Rat
Clone: MF23
Isotype: IgG2b
Species Reactivity: Mouse
Immunogen: Mouse Foxp3 Recombinant Protein

Intracellular Staining

Spezifikation

Spezifikation

Foxp3
Monoclonal
0.2mg/mL
Forkhead box p3; Forkhead box protein p3; JM2; Scurfin; Scurfy; Sf
Rat
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
MF23
Alexa Fluor 488
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse Foxp3 Recombinant Protein
25 μg
Immunology
Mouse
IgG2b
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.