missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-Foxp3; Clone: R16-715; BD
Marke: BD Biosciences 563101
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Host Species: Rat
Clone R16-715
Isotype IgG2a, κ
Species Reactivity Ms
Immunogen Recombinant Protein
Flow Cytometry, Intracellular Staining
Spezifikation
Foxp3 | |
R16-715 | |
PE | |
Recombinant Protein | |
100 μg | |
Primary | |
IgG2a |
Monoclonal | |
0.2mg/mL | |
Rat | |
Affinity Purified | |
RUO | |
Mouse |