missing translation for 'onlineSavingsMsg'
Learn More

Frizzled-1 Rat anti-Human, Mouse, Clone: 162531, R&D Systems™

Rat Monoclonal Antibody has been used in 1 publication

N.V.

Spezifikation

Antigen Frizzled-1
Klon 162531
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-1 Gln72-His248 (Met122Ile) Accession # O70421
Wirtsspezies Rat
Primär oder sekundär Primary
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

Frizzled-1 Monoclonal specifically detects Frizzled-1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Spezifikation

Spezifikation

Frizzled-1
Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-1 Gln72-His248 (Met122Ile) Accession # O70421
Primary
Human, Mouse
Unconjugated
8321
Purified
Western Blot, Flow Cytometry, CyTOF
RUO
162531
Rat
Monoclonal
IgG2a
DKFZp564G072, FLJ95923, frizzled family receptor 1, frizzled homolog 1, frizzled, Drosophila, homolog of, 1, frizzled-1, Frizzled1, fz-1, FZD1, fzE1, hFz1, seven transmembrane spanning receptor, Wnt receptor
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen