Learn More
Frizzled-1 Rat anti-Human, Mouse, Clone: 162531, R&D Systems™
Rat Monoclonal Antibody
Marke:  RD Systems (Bio-Techne) MAB11201-050
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13418365
  • 187.44 € / 50 Mikrogramm
Voraussichtlicher Liefertermin 30-07-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Frizzled-1 Monoclonal antibody MAB11201SP Specifically detects Frizzled-1 in Human, Mouse samples. It is validated for Flow Cytometry, Western Blotting
Spezifikation
Spezifikation
Frizzled-1
162531
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
FZD1
Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-1 Gln72-His248 (Met122Ile) Accession # O70421
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
8321
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Flow Cytometry, CyTOF
LYOPH
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
O70421
DKFZp564G072, FLJ95923, frizzled family receptor 1, frizzled homolog 1, frizzled, Drosophila, homolog of, 1, frizzled-1, Frizzled1, fz-1, FZD1, fzE1, hFz1, seven transmembrane spanning receptor, Wnt receptor
Rat
IgG2a
50 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human and mouse Frizzled-1 in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant mouse (rm) Frizzled-9 is observed and no cross-reactivity with recombinant human (rh) Frizzled-5, rmFrizzled 2, 3, 4, 6, 7, 8, or rhMFRP is observed.
Human, Mouse
Videos