missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frizzled-2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518884
Weitere Details : Netto-Gewicht : 0.01000kg
Descripción
This antibody is predicted to react with mouse and rat based on sequence homology.
Frizzled-2 Polyclonal antibody specifically detects Frizzled-2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot.Especificaciones
Frizzled-2 | |
Polyclonal | |
Unconjugated | |
Fzd2 | |
AL033370; AW456835; Dfrizzled2; Drosophila polarity gene (frizzled) homologue; Drosophila polarity gene homolog 2; frizzled 2, seven transmembrane spanning receptor; frizzled class receptor 2; frizzled family receptor 2; frizzled homolog 2; frizzled homolog 2 (Drosophila); frizzled-10; Frizzled-2; Fz10; fz-10; Fz2; Fz-2; Fzd10; Fzd2; fzE2; hFz2; Mfz10; Mfz10a; mFz2; rFz2 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
2535 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q14332 | |
Fzd2 | |
Synthetic peptide sequence (RYATLEHPFHCPR) corresponding to the internal amino acids of FZD2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |