missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fumarase Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518745
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with canine, mouse and rat based on sequence homology.
Fumarase Polyclonal antibody specifically detects Fumarase in Human samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Spezifikation
Fumarase | |
Polyclonal | |
Unconjugated | |
FH | |
EF-3; Fh; Fh1; Fh-1; FMRD; fumarase; fumarate hydratase; fumarate hydratase 1; fumarate hydratase, mitochondrial; HLRCC; LRCC; MCL; MCUL1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
2271 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P07954 | |
FH | |
Synthetic peptide sequence (HPNDHVNKSQSSND) corresponding to the internal amino acids of FH | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |