missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma Catenin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA517320
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with -beta-catenin.
gamma Catenin Polyclonal antibody specifically detects gamma Catenin in Hamster, Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Spezifikation
gamma Catenin | |
Polyclonal | |
Unconjugated | |
JUP | |
ARVD12; catenin (cadherin-associated protein), gamma 80kDa; Catenin gamma; cb80; cell-adhesion protein plakoglobin; Ctnng; D930025P04Rik; danPG; Desmoplakin III; desmoplakin-3; desmosomal protein; DP3; DPIII; fb11b06; gama-catenin; gamma-catenin; gamma-catenin (plakoglobin); junction plakoglobin; junction plakoglobin a; Jup; jup protein; jupa; OTTHUMP00000164735; PDGB; PG; PKGB; plakoglobin; wu:fb11b06 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100760007, 16480, 3728, 81679 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
32 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P14923, Q02257, Q6P0K8 | |
JUP | |
Synthetic peptide corresponding to the carboxy terminus of human gamma-catenin | |
100 μL | |
Primary | |
Hamster, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |