Learn More
gamma Tubulin Monoclonal Antibody (TU-30), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA119421
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody recognizes the C-terminal peptide sequence EYHAATRPDYISWGTQ (aa 434-449) of gamma-tubulin. Immunocytochemistry: Staining technique: (a) Fix cells for 10 min in methanol at -20°C and for 6 min in acetone at -20°C; (b) Fix cells directly in methanol for 10 min at -20°C or in acetone for 10 min at -20°C. The antibody TU-30 stains only fixed cells; Western Blot: Reducing conditions.
gamma Tubulin Monoclonal antibody specifically detects gamma Tubulin in Bovine, Chicken, Human, Mouse, Plant, Porcine, Protozoa, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.Spezifikation
gamma Tubulin | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P23258, P83887, P83888, Q0VCD2, Q32KM1, Q8VCK3, Q9NRH3 | |
TUBG1, Tubg2 | |
C-terminal peptide of gamma-Tubulin counjugated to KLH. | |
100 μg | |
Primary | |
Bovine, Chicken, Human, Mouse, Plant, Porcine, Protozoa, Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Western Blot | |
TU-30 | |
Unconjugated | |
TUBG1 | |
1500010O08Rik; AI451582; AI503389; AI504772; CDCBM4; centrosome marker; gamma tubulin; gamma-1-tubulin; Gamma-2-tubulin; gamma-tubulin; gamma-tubulin complex component 1; GCP-1; similar to tubulin, gamma 1; Tubg; TUBG1; tubg1.L; TUBG2; TUBGCP1; Tubgl; tubulin gamma 1; tubulin gamma 1 L homeolog; tubulin gamma 2; Tubulin gamma-1 chain; Tubulin gamma-2 chain; tubulin, gamma 1; tubulin, gamma 2; tubulin, gamma polypeptide; Unknown (protein for MGC:133882); XELAEV_18002398mg; xgam; zgc:55894 | |
Mouse | |
Protein A | |
RUO | |
100520862, 103733, 103768, 106505426, 252921, 27175, 420025, 540216, 541101, 680991, 7283 | |
4° C, do not freeze | |
Liquid |