missing translation for 'onlineSavingsMsg'
Learn More

GCDH Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA530040

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13464487

  • 473.00 € / 100 Mikroliter
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Recommended positive controls: HepG2. Predicted reactivity: Mouse (88%), Rat (89%), Xenopus laevis (84%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

GCDH Polyclonal antibody specifically detects GCDH in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Spezifikation

Spezifikation

GCDH
Polyclonal
Unconjugated
GCDH
9030411L18; ACAD5; AI266902; D17825; GCD; GCDH; glutaryl-CoA dehydrogenase; glutaryl-CoA dehydrogenase, mitochondrial; glutaryl-Coenzyme A dehydrogenase; MGC137369 protein
Rabbit
Antigen affinity chromatography
RUO
2639, 270076
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q60759, Q92947
GCDH
Recombinant fragment corresponding to a region within amino acids 9 and 229 of Human GCD
100 μL
Primary
Human, Mouse
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote