missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYF2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518108
Weitere Details : Netto-Gewicht : 0.01000kg
Description
This antibody is predicted to react with bovine, canine and rat based on sequence homology.
SYF2 Polyclonal antibody specifically detects SYF2 in Human, Mouse samples. It is validated for Western Blot.Spécification
SYF2 | |
Polyclonal | |
Unconjugated | |
SYF2 | |
1110018L13Rik; Cbpin; CCNDBP1 interactor; CCNDBP1-interactor; D4Bwg1551e; DKFZP564O2082; fSAP29; functional spliceosome-associated protein 29; GCIP-interacting protein p29; GCIP-interacting protein p29 (Gcipip); Gcipip; mp29; Ntc31; P29; Pre-mRNA-splicing factor SYF2; SYF2; SYF2 homolog, RNA splicing factor; SYF2 homolog, RNA splicing factor (S. cerevisiae); SYF2 pre-mRNA splicing factor; SYF2 pre-mRNA-splicing factor | |
Goat | |
Antigen affinity chromatography | |
RUO | |
25949, 68592 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O95926, Q9D198 | |
SYF2 | |
Synthetic peptide sequence (AEIKQNLERGTAV) corresponding to the C-terminus amino acids of SYF2 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |