Learn More
GCLM Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529908
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HeLa, A431, mouse liver, rat liver, rat kidney. Predicted reactivity: Mouse (97%), Rat (96%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GCLM Polyclonal antibody specifically detects GCLM in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
GCLM | |
Polyclonal | |
Unconjugated | |
Gclm | |
AI649393; gamma glutamylcysteine synthetase; gamma-ECS regulatory subunit; Gamma-glutamylcysteine synthetase regulatory subunit; Gclm; Gcmc; GCS light chain; Glclr; glutamate cysteine ligase (gamma-glutamylcysteine synthetase), regulatory; glutamate cysteine ligase, modifier subunit; glutamate-cysteine ligase (gamma-glutamylcysteine synthetase), regulatory (30.8kD); glutamate-cysteine ligase modifier subunit; glutamate--cysteine ligase modifier subunit; glutamate-cysteine ligase modifier subunit delta2 alternative splicing; glutamate-cysteine ligase regulatory protein; Glutamate--cysteine ligase regulatory subunit; glutamate-cysteine ligase, modifier subunit; GSC light chain; null | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14630, 2730, 29739 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.33 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O09172, P48507, P48508 | |
Gclm | |
Recombinant fragment corresponding to a region within amino acids 1 and 261 of Human GCLM | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |