Learn More
GCM1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529438
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HepG2, HepG2 nucleus. Predicted reactivity: Mouse (84%), Rat (84%), Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GCM1 Polyclonal antibody specifically detects GCM1 in Human samples. It is validated for Western Blot.Spezifikation
GCM1 | |
Polyclonal | |
Unconjugated | |
GCM1 | |
chorion-specific transcription factor GCMa; Gcm 1; Gcm a; GCM motif protein 1; GCM1; Gcm1-rs1; Gcma; Gcm-rs2; glial cell deficient; glial cells missing homolog 1; glial cells missing homolog 1 (Drosophila); glial cells missing transcription factor 1; glide; hGCMa; mGCM1; mGCMa | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8521 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.84 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9NP62 | |
GCM1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 277 of Human GCM1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |