Learn More
GCNF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527290
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: NT2D1. Predicted reactivity: Mouse (100%), Xenopus laevis (80%), Pig (100%), Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GCNF Polyclonal antibody specifically detects GCNF in Human samples. It is validated for Western Blot.Spezifikation
GCNF | |
Polyclonal | |
Unconjugated | |
NR6A1 | |
1700113M01Rik; CT150; Gcnf; GCNF1; Germ cell nuclear factor; germ cell nuclear factor variant 1; h hRTR; hGCNF; hRTR; mGCNF; NCNF; NR61; NR6A1; Nuclear receptor subfamily 6 group A member 1; nuclear receptor subfamily 6, group A, member 1; retinoic acid receptor-related testis-associated receptor; retinoid receptor-related testis-specific receptor; RTR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2649 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.76 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q15406 | |
NR6A1 | |
Recombinant fragment corresponding to a region within amino acids 303 and 465 of Human GCNF | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |