missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCNT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA512146
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
GCNT1 Polyclonal antibody specifically detects GCNT1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
GCNT1 | |
Polyclonal | |
Unconjugated | |
GCNT1 | |
5630400D21Rik; B130048E03; beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase; beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N--acetylglucosaminyltransferase; beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase (Core 2 branching enzyme) (Core2-GlcNAc-transferase) (C2GNT); beta1,6 N-acetylglucosaminyltransferase; beta-1,6-N-acetylglucosaminyltransferase; C2 GlcNAcT; C2GNT; C2GNT1; C2GNT-L; core 2 beta1,6 N-acetylglucosaminyltransferase-I; core 2 beta-1,6-N-acetylglucosaminyltransferase I; core 2 branching enzyme; core 2 GlcNAc-T; core 2 GnT; core 2-branching enzyme; Core2-GlcNAc-transferase; enzymatic glycosylation-regulating; G6NT; Gcnt1; glucosaminyl (N-acetyl) transferase 1, core 2; glucosaminyl (N-acetyl) transferase 1, core 2 (beta-1,6-N-acetylglucosaminyltransferase); glucosaminyl transferase 1, core 2; IGnT; NACGT2; NAGCT2 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
2650 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
Q02742 | |
GCNT1 | |
KLH conjugated synthetic peptide between 399-428 amino acids from the C-terminal region of human GCNT1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |