Learn More
GCP4 Monoclonal Antibody (GT812), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA517294
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HeLa. Predicted reactivity: Mouse (97%), Zebrafish (90%), Xenopus laevis (90%), Rabbit (97%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GCP4 Monoclonal antibody specifically detects GCP4 in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
GCP4 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q9UGJ1 | |
TUBGCP4 | |
Recombinant protein fragment contains a sequence corresponding to a region within amino acids 1 and 178 of GCP4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunocytochemistry, Western Blot | |
GT812 | |
Unconjugated | |
TUBGCP4 | |
4932441P04Rik; 76P; D2Ertd435e; gamma tubulin ring complex protein; gamma tubulin ring complex protein (76p gene); gamma-ring complex protein 76 kDa; Gamma-tubulin complex component 4; GCP4; GCP-4; Grip76; h76p; hGCP4; hGrip76; MCCRP3; RGD1306924; TUBGCP4; tubulin gamma complex associated protein 4; tubulin, gamma complex associated protein 4 | |
Mouse | |
Protein G | |
RUO | |
27229 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |