missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMA Monoclonal Antibody (GCP-05), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA110335
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody recognizes an extracellular domain, preferentially in native form, of glutamate carboxypeptidase II. It has also been sucessfully used in Mass Cytometry (CyTof).
PSMA Monoclonal antibody specifically detects PSMA in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunoprecipitation.Spezifikation
PSMA | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
Q04609 | |
FOLH1 | |
amino acids 44-750 of human GCPII | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunoprecipitation | |
GCP-05 | |
Unconjugated | |
FOLH1 | |
Cell growth-inhibiting gene 27 protein; FGCP; folate hydrolase (prostate-specific membrane antigen) 1; folate hydrolase 1; FOLH; Folh1; Folylpoly-gamma-glutamate carboxypeptidase; GCP2; GCPII; GIG27; glutamate carboxylase II; glutamate carboxypeptidase 2; Glutamate carboxypeptidase II; membrane glutamate carboxypeptidase; mGCP; mopsm; Naalad; NAALAD1; NAALAdase; NAALADase I; N-acetylated alpha-linked acidic dipeptidase; N-acetylated alpha-linked acidic dipeptidase 1; N-acetylated-alpha-linked acidic dipeptidase I; prostate specific membrane antigen; prostate specific membrane antigen variant F; prostate-specific membrane antigen; Prostate-specific membrane antigen homolog; PSM; PSMA; pteroylpoly-gamma-glutamate carboxypeptidase | |
Mouse | |
Protein A | |
RUO | |
2346 | |
4° C, do not freeze | |
Liquid |