missing translation for 'onlineSavingsMsg'
Learn More

GDNF Polyclonal Antibody, Invitrogen™

Goat Polyclonal Antibody

Marke:  Invitrogen PA124827

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11555523

  • 514.00 € / 200 Mikrogramm
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Neutralization: The ND50 is 2.6 μg/mL. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

GDNF Polyclonal antibody specifically detects GDNF in Human samples. It is validated for Inhibition Assays, Neutralization.
Spezifikation

Spezifikation

GDNF
Polyclonal
Unconjugated
GDNF
AI385739; Astrocyte-derived trophic factor; ATF; ATF1; ATF2; Gdnf; glial cell derived neurotrophic factor; glial cell line derived neurotrophic factor; Glial cell line derived neutrophic factor; glial cell line-derived neurotrophic factor; gndf; HFB1-GDNF; hGDNF; H-GDNF; HSCR3; mGDNF; neurotrophic factor
Goat
Protein G
RUO
2668
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Inhibition Assays, Neutralization
1 mg/mL
PBS with no preservative
P39905
GDNF
Recombinant, full length human GDNF protein expressed in E. coli.
200 μg
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote