missing translation for 'onlineSavingsMsg'
Learn More

GEF-H1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA532213

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13426737

  • 534.00 € / 100 Mikroliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Recommended positive controls: 293T, A431, BCL-1, C2C12, C8D30, GL261, HeLa, HepG2, NIH-3T3, Neuro 2A, Raw264.7, hMSC-3A6, PC-12, Rat2, mouse fibroblast. Predicted reactivity: Mouse (89%), Rat (89%), Dog (92%), Pig (95%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

GEF-H1 Polyclonal antibody specifically detects GEF-H1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.
Spezifikation

Spezifikation

GEF-H1
Polyclonal
Unconjugated
ARHGEF2
AA408978; ARHGEF2; CG10188; CG10188-PA; CG10188-PB; CG10188-PC; cyst; cyst-PA; cyst-PB; cyst-PC; cysts; DKFZp547L106; DKFZp547P1516; Dmel\CG10188; Dmel_CG10188; FLJ45440; GEF; GEFH1; GEF-H1; Guanine nucleotide exchange factor H1; Kiaa0651; Lbcl1; LBC'S first cousin; Lfc; LFP40; Lymphoid blast crisis-like 1; microtubule-regulated Rho-GEF; mKIAA0651; oncogene LFC; OTTHUMP00000018855; P40; Proliferating cell nucleolar antigen p40; rho guanine nucleotide exchange factor 2; rho guanine nucleotide exchange factor 2-like protein; Rho/Rac guanine nucleotide exchange factor (GEF) 2; Rho/Rac guanine nucleotide exchange factor 2; rhobin
Rabbit
Antigen affinity chromatography
RUO
16800, 310635, 9181
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.51 mg/mL
PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Q5FVC2, Q60875, Q92974
ARHGEF2
Recombinant fragment corresponding to a region within amino acids 656 and 986 of Human GEF-H1
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote