Learn More
GEFT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531827
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7, mouse brain. Predicted reactivity: Mouse (88%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GEFT Polyclonal antibody specifically detects GEFT in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
GEFT | |
Polyclonal | |
Unconjugated | |
Arhgef25 | |
2410008H17Rik; ARHGEF25; D10Ertd610e; Geft; guanine nucleotide exchange factor GEFT; p63RhoGEF; RAC/CDC42 exchange factor; RAC/CDC42 exchange factor isoform 2; rac/Cdc42/Rho exchange factor GEFT; Rho guanine nucleotide exchange factor (GEF) 25; Rho guanine nucleotide exchange factor 25; RhoA/RAC/CDC42 exchange factor; rhoA/Rac/Cdc42 guanine nucleotide exchange factor GEFT | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
115557, 52666 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q86VW2, Q9CWR0 | |
Arhgef25 | |
Recombinant protein fragment corresponding to a region within amino acids 226 and 446 of Human GEFT | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |