Learn More
Gelsolin Mouse anti-Human, Clone: 35B2, Invitrogen™
Mouse Monoclonal Antibody
Marke:  Invitrogen LFMA0176
Weitere Details : Netto-Gewicht : 0.99000kg
Artikelnummer. 10318569
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Beschreibung
Beschreibung
A suggested positive control for this product is human plasma.
The protein encoded by this gene binds to the "plus" ends of actin monomers and filaments to prevent monomer exchange. The encoded calcium-regulated protein functions in both assembly and disassembly of actin filaments. Defects in this gene are a cause of familial amyloidosis Finnish type (FAF). Multiple transcript variants encoding several different isoforms have been found for this gene.
Spezifikation
Spezifikation
Gelsolin
Monoclonal
Unconjugated
P06396
GSN
Protein purified from Human plasma
100 μL
Primary
Human
Antibody
IgG1
Immunoprecipitation, Western Blot
35B2
GSN
RP11-477J21.1, DKFZp313L0718
Mouse
Protein G
RUO
2934
-20° C, Avoid Freeze/Thaw Cycles