missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gelsolin Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518605
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with canine, mouse and rat based on sequence homology.
Gelsolin Polyclonal antibody specifically detects Gelsolin in Human samples. It is validated for Flow Cytometry, Western Blot.Spezifikation
Gelsolin | |
Polyclonal | |
Unconjugated | |
GSN | |
Actin-depolymerizing factor; ADF; ADFBrevin; AGEL; Brevin; DKFZp313L0718; gelsolin; gelsolin (amyloidosis, Finnish type); Gsb; Gsn; hypothetical protein LOC535077; plasma gelsolin precursor; RP11-477J21.1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
2934 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P06396 | |
GSN | |
Synthetic peptide sequence (PRLKDKKMDAHP) corresponding to the internal amino acids of GSN | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |