missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gemin 7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA546733
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: MQTPVNIPVP VLRLPRGPDG FSRGFAPDGR RAPLRPEVPE IQECPIAQES Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rat: 100%
Gemin 7 Polyclonal antibody specifically detects Gemin 7 in Human samples. It is validated for Western Blot.Spezifikation
Gemin 7 | |
Polyclonal | |
Unconjugated | |
Gemin7 | |
2400008I04Rik; AI120175; AI461688; gem (nuclear organelle) associated protein 7; gem nuclear organelle associated protein 7; gem-associated protein 7; gemin 7; Gemin7; gemin-7; KIAA2024; SIP3; ZNF650 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
79760 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H840 | |
Gemin7 | |
Synthetic peptide directed towards the N-terminal of human GEMIN7 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |