missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gephyrin Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA519589
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with chicken, human and Xenopus laevis based on sequence homology.
This gene encodes a neuronal assembly protein that anchors inhibitory neurotransmitter receptors to the postsynaptic cytoskeleton via high affinity binding to a receptor subunit domain and tubulin dimers. In nonneuronal tissues, the encoded protein is also required for molybdenum cofactor biosynthesis. Mutations in this gene may be associated with the neurological condition hyperplexia and also lead to molybdenum cofactor deficiency. Numerous alternatively spliced transcript variants encoding different isoforms have been described; however, the full-length nature of all transcript variants is not currently known.Spezifikation
Gephyrin | |
Polyclonal | |
Unconjugated | |
Q8BUV3, Q03555 | |
GPHN | |
Synthetic peptide conjugated to KLH derived from within residues 700 to the C-terminus of Mouse Gephyrin. | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.4 mg/mL | |
Gphn | |
GEPH, GPH, GPHRYN, KIAA1385 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
268566, 64845 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |