missing translation for 'onlineSavingsMsg'
Learn More

TRIM8 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA531428

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13495947

  • 514.00 € / 100 Mikroliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Recommended positive controls: HepG2, 3XFlag-human TRIM8-transfected 293T. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (91%), Cat (100%), Chicken (94%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TRIM8 Polyclonal antibody specifically detects TRIM8 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
Spezifikation

Spezifikation

TRIM8
Polyclonal
Unconjugated
TRIM8
AA408830; BB006755; E3 ubiquitin-protein ligase TRIM8; GERP; glioblastoma expressed ring finger protein; glioblastoma-expressed RING finger protein; Probable E3 ubiquitin-protein ligase TRIM8; ring finger protein 27; RING-type E3 ubiquitin transferase TRIM8; Rnf27; TRIM8; tripartite motif containing 8; tripartite motif protein 8; tripartite motif protein TRIM8; tripartite motif-containing 8; tripartite motif-containing protein 8; tripartite motif-containing protein 8; probable E3 ubiquitin-protein ligase TRIM8
Rabbit
Antigen affinity chromatography
RUO
81603
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Q9BZR9
TRIM8
Recombinant fragment corresponding to a region within amino acids 17 and 242 of Human GERP
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote