missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFAP Monoclonal Antibody (6F2), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA135377
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody stains the 56 kDa GFAP band in immunoblots performed on crude brain extracts. It shows no cross-reactivity to Keratin, Vimentin or Neurofilament.
GFAP Monoclonal antibody specifically detects GFAP in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Spezifikation
GFAP | |
Monoclonal | |
25 μg/mL | |
tissue culture supernatant with 0.01M sodium azide | |
P03995, P14136 | |
GFAP | |
GFAP from human brain. | |
RUO | |
14580, 2670 | |
4° C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
6F2 | |
Unconjugated | |
GFAP | |
6330404F12Rik; 65 kDa glutamic acid decarboxylase; AI836096; ALXDRD; Astrocyte or Intermediate Filament Protein; cb345; class III intermediate filament protein; etID36982.3; FLJ45472; GAD(65); Gad2; Gad-2; GAD65; GAD-65; GFAP; GFAP epsilon; gfap protein; gfapl; glial fibrillary acidic protein; Glial Fibrillary Acidic Protein (GFAP); glial fibrillary acidic protein alpha; glutamate decarboxylase 2; Glutamate decarboxylase 65 kDa isoform; glutamic acid decarboxylase 2; I79_011607; intermediate filament; intermediate filament protein; intermediate filament protein; GFAP; Unknown (protein for MGC:139638); wu:fb34h11; wu:fk42c12; xx:af506734; zgc:110485; zGFAP; intermediate filament protein; zrf-1 antigen | |
Mouse | |
1 mL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |