Learn More
GFAP Monoclonal Antibody (GF-01), Invitrogen™
Mouse Monoclonal Antibody
Marke:  Invitrogen MA119170
Weitere Details : Netto-Gewicht : 0.01000kg
Artikelnummer. 11594532
  • 422.00 € / 100 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
This antibody will not cross-react with rat.
GFAP Monoclonal antibody specifically detects GFAP in Feline, Human, Porcine samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.
Spezifikation
Spezifikation
GFAP
Monoclonal
1 mg/mL
PBS with 15mM sodium azide; pH 7.4
P14136
GFAP
Pellet of pig brain cold-stable proteins after depolymerization of Microtubules.
100 μg
Primary
Feline, Human, Porcine
Antibody
IgG1
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
GF-01
Unconjugated
GFAP
6330404F12Rik; 65 kDa glutamic acid decarboxylase; AI836096; ALXDRD; Astrocyte or Intermediate Filament Protein; cb345; class III intermediate filament protein; etID36982.3; FLJ45472; GAD(65); Gad2; Gad-2; GAD65; GAD-65; GFAP; GFAP epsilon; gfap protein; gfapl; glial fibrillary acidic protein; Glial Fibrillary Acidic Protein (GFAP); glial fibrillary acidic protein alpha; glutamate decarboxylase 2; Glutamate decarboxylase 65 kDa isoform; glutamic acid decarboxylase 2; I79_011607; intermediate filament; intermediate filament protein; intermediate filament protein; GFAP; Unknown (protein for MGC:139638); wu:fb34h11; wu:fk42c12; xx:af506734; zgc:110485; zGFAP; intermediate filament protein; zrf-1 antigen
Mouse
Protein A
RUO
101081938, 2670, 396562
4° C, do not freeze